Thermo Scientific Chemicals

Sodium chlorodifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: A15756.14
25 g, Each
Thermo Scientific Chemicals

Sodium chlorodifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: A15756.14
25 g, Each
Quantity
Catalog number: A15756.14
also known as A15756-14
Price (USD)
Price: 79.20
Online price: 67.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1895-39-2
IUPAC Name
sodium 2-chloro-2,2-difluoroacetate
Molecular Formula
C2ClF2NaO2
InChI Key
MRTAVLDNYYEJHK-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C(F)(F)Cl
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

Solubility
Soluble in dimethoxyethane, diglyme (0.60 g mL-1), triglyme, dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text