Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
- E. Hadjoudis.; T. Dziembowska.; Z. Rozwadowski. Photoactivation of the thermochromic solid di-anil of 2-hydroxy-5-methyl-isophthalaldehyde in β-cyclodextrin..J. Photochem. Photobiol., A 1999, 128 (1-3), 97-99 .
- Jürgen Lorösch.; Wolfgang Haase. Crystal structure and magnetic properties of an asymmetric bridged binuclear Cu(II)-complex with a macrocyclic ligand derived from 2-hydroxy-5-methyl-isophthalaldehyde and histamine.Inorg. Chim. Acta. 1985, 108 (1),35-40 .