3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
Thermo Scientific Chemicals

3-Nitrobenzonitrile, 98%

CAS: 619-24-9 | C7H4N2O2 | 148.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
250 g
Catalog number A15821.14
also known as A15821-14
Price (EUR)
53,80
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
53,80
Each
Chemical Identifiers
CAS619-24-9
IUPAC Name3-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeyRUSAWEHOGCWOPG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)113.0-119.0?C
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or lumps or needles
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only