Thermo Scientific Chemicals

4-Bromotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A15843.22
100 g, Each
Thermo Scientific Chemicals

4-Bromotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A15843.22
100 g, Each
Quantity
Catalog number: A15843.22
also known as A15843-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
106-38-7
IUPAC Name
1-bromo-4-methylbenzene
Molecular Formula
C7H7Br
InChI Key
ZBTMRBYMKUEVEU-UHFFFAOYSA-N
SMILES
CC1=CC=C(Br)C=C1
Form
Low-melting fused solid (clear liquid as melt)
Assay (GC)
≥97.5%
Appearance (Color)
White

Description

4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text