3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%

CAS: 1620-98-0 | C15H22O2 | 234.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15864.14
also known as A15864-14
Price (EUR)
97,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
97,00
Each
Chemical Identifiers
CAS1620-98-0
IUPAC Name3,5-di-tert-butyl-4-hydroxybenzaldehyde
Molecular FormulaC15H22O2
InChI KeyDOZRDZLFLOODMB-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(C=O)=CC(=C1O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to yellow
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)185-193°C
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

Solubility
Soluble in hot methanol.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Michael D. Mullican.; Michael W. Wilson.; David T. Conner.; Catherine R. Kostlan.; Denis J. Schrier.; Richard D. Dyer. Design of 5-(3,5-di-tert-butyl-4-hydroxyphenyl)-1,3,4-thiadiazoles, -1,3,4-oxadiazoles, and -1,2,4-triazoles as orally active, nonulcerogenic antiinflammatory agents.J. Med. Chem. 1993, 36 (8),1090-1099 .