1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
Thermo Scientific Chemicals

1,8-Diaminonaphthalene, 97%

CAS: 479-27-6 | C10H10N2 | 158.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A15966.0B
also known as A15966-0B
Price (USD)
646.65
Online Exclusive
718.00
Save 71.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
646.65
Online Exclusive
718.00
Save 71.35 (10%)
Each
Chemical Identifiers
CAS479-27-6
IUPAC Namenaphthalene-1,8-diamine
Molecular FormulaC10H10N2
InChI KeyYFOOEYJGMMJJLS-UHFFFAOYSA-N
SMILESNC1=CC=CC2=CC=CC(N)=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown or purple
Assay (GC)≥96.0%
Identification (FTIR)Conforms
FormCrystals, powder, crystalline lumps/powder or flakes
Melting Point60.0-68.0?C
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Xin-Gui Li.; Mei-Rong Huang.; Sheng-Xian Li. Facile synthesis of poly(1,8-diaminonaphthalene) microparticles with a very high silver-ion adsorbability by a chemical oxidative polymerization. Acta Materialia. 2004, 52 (18), 5363-5374 .
  2. B.J. Palys.; J. Bukowska.; K. Jackowska. SERS of 1,8-diaminonaphthalene on gold, silver and copper electrodes Polymerisation and complexes formed with the electrode material. Journal of Electroanalytical Chemistry. 1997, 428 (1-2), 19-24.