Thermo Scientific Chemicals

1-Octanol, 99%, Thermo Scientific Chemicals

Catalog number: A15977.0F
2500 mL, Each
Thermo Scientific Chemicals

1-Octanol, 99%, Thermo Scientific Chemicals

Catalog number: A15977.0F
2500 mL, Each
Quantity
Catalog number: A15977.0F
also known as A15977-0F
Price (USD)
Price: 120.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-87-5
IUPAC Name
octan-1-ol
Molecular Formula
C8H18O
InChI Key
KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES
CCCCCCCCO
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4280-1.4310 @ 20?C
Form
Liquid
Assay (GC)
≥98.5%

Description

1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text