4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals
4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals
4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals
4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A16029.14
also known as A16029-14
25 g
Catalog number A16029.14
also known as A16029-14
Price (EUR)
414,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
414,00
Each
Add to cart
Chemical Identifiers
CAS348-60-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or chunks or flakes
Melting Point (clear melt)50.0-56.0?C
Appearance (Color)White to cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only