4-Phenoxyaniline, 97%
4-Phenoxyaniline, 97%
4-Phenoxyaniline, 97%
4-Phenoxyaniline, 97%
Thermo Scientific Chemicals

4-Phenoxyaniline, 97%

CAS: 139-59-3 | C12H11NO | 185.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16048.14
also known as A16048-14
Price (EUR)
34,30
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
34,30
Each
Add to cart
Chemical Identifiers
CAS139-59-3
IUPAC Name4-phenoxyaniline
Molecular FormulaC12H11NO
InChI KeyWOYZXEVUWXQVNV-UHFFFAOYSA-N
SMILESNC1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)80.0-89.0?C
Appearance (Color)Pale cream to cream to dark brown to brown
FormPowder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only