Thermo Scientific Chemicals

Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A16084.AU
1000 mL, Each
Thermo Scientific Chemicals

Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A16084.AU
1000 mL, Each
Quantity

Chemical Identifiers

CAS
126-73-8
IUPAC Name
tributyl phosphate
Molecular Formula
C12H27O4P
InChI Key
STCOOQWBFONSKY-UHFFFAOYSA-N
SMILES
CCCCOP(=O)(OCCCC)OCCCC
Form
Liquid
Assay from Supplier's CofA
≥97.5% (US sourced material)
Appearance (Color)
Clear colorless
Color (APHA)
≤50 (US sourced material)
Comment
Material sourced in UK or US

Description

Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

Solubility
Miscible with diethyl ether, benzene, carbon disulfide and ethanol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text