Acetamidine hydrochloride, 97%
Acetamidine hydrochloride, 97%
Acetamidine hydrochloride, 97%
Thermo Scientific Chemicals

Acetamidine hydrochloride, 97%

CAS: 124-42-5 | C2H7ClN2 | 94.542 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16091.14
also known as A16091-14
Price (EUR)
22,70
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
22,70
Each
Chemical Identifiers
CAS124-42-5
IUPAC Namehydrogen ethanimidamide chloride
Molecular FormulaC2H7ClN2
InChI KeyWCQOBLXWLRDEQA-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or chunks
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to cream
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

Solubility
Soluble in water.

Notes
It is hygroscopic. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Michael Schnoor; Peter Voss; Paul Cullen; Thomas Boking; Hans-Joachim Galla; Erwin A.Galinski; Stefan Lorkowski. Characterization of the synthetic compatible solute homoectoine as a potent PCR enhancer. Biochemical and Biophysical Research Communications. 2004, 322, (3),867-872
  2. H.Niwa; S.Inouye; T Hirano; T.Matsuno; S.Kojima; M.Kubota; M.Ohashi; F.I.Tsuji. Chemical nature of the light emitter of the Aequorea green fluorescent protein. PNAS. 1996, 93, (24),13617-13622