Invitrogen™

Goat anti-Rabbit IgG (H+L) - cross-adsorbed, horseradish peroxidase (HRP) conjugate

Catalog number: A16104SAMPLE
Invitrogen™

Goat anti-Rabbit IgG (H+L) - cross-adsorbed, horseradish peroxidase (HRP) conjugate

Catalog number: A16104SAMPLE
Catalog Number
A16104SAMPLE
Unit Size
50 µg
Price (USD)
64.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A16104SAMPLE50 µg
64.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
This product is a sample size (50 µG) of Goat anti-Rabbit IgG (H+L) - cross-adsorbed, horseradish peroxidase (HRP) Conjugate, (A16104).

Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, is prepared from antibodies that have been adsorbed against bovine, human, and mouse IgG to minimize cross reactivity. Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, should be suitable for western blot, ELISA, immunohistochemistry and other standard immunoassay applications. The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by isoelectric focusing (IEF).

Applications: Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, should be suitable for western blot, ELISA, immunohistochemistry and other standard immunoassay applications.
Host species: The host species of theanti-rabbit IgG (H+L), horseradish peroxidase (HRP) Conjugate, is goat.
Reactivity: Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, detects rabbit IgG (H+L).
Product size: 50 µg
Suggested dilution: The optimal working dilution should be determined empirically. Dilutions of 1:500-1:5000 should be satisfactory for most immunohistochemical applications. Dilutions of 1:200-1:5,000 should be satisfactory for most ELISA and western blot applications.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Applications
Immunoassay
Conjugate
HRP
Description
Goat anti-Rabbit IgG (H+L) - Cross-adsorbed, 50μg, Lyophilized, Goat Host, Mouse Species Reactivity, HRP Conjugate, IgG Isotype, 2 to 8°C Storage Temperature, For Cellular Imaging, Protein Assays and Analysis, Secondary Detection
Format
Lyophilized
Host Species
Goat
Isotype
IgG
Purification Method
Affinity Purified
Quantity
50μg
Regulatory Status
RUO
Storage Requirements
2 to 8°C
Type
Secondary
Primary or Secondary
Secondary
Packaging
1 Each
Target Species
Murine
Antibody Conjugate
Enzyme Labeled, HRP (Horseradish Peroxidase)
Antibody Fragment
Whole Antibody
Form
Lyophilized
Label or Dye
HRP (Horseradish Peroxidase)
Label Type
Enzyme Labeled
Reactivity
Mouse
Target Isotype
IgG

Contents & Storage

Store at 2–8°C. For long term storage of antibody solutions, dilute with 50% glycerol, aliquot, and store at -5 to -35°C.

Figures

Frequently asked questions (FAQs)

Citations & References