5-Bromo-2-fluorobenzonitrile, 98%
5-Bromo-2-fluorobenzonitrile, 98%
5-Bromo-2-fluorobenzonitrile, 98%
5-Bromo-2-fluorobenzonitrile, 98%
Thermo Scientific Chemicals

5-Bromo-2-fluorobenzonitrile, 98%

CAS: 179897-89-3 | C7H3BrFN | 200.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16113.14
also known as A16113-14
Price (EUR)
91,80
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
91,80
Each
Add to cart
Chemical Identifiers
CAS179897-89-3
IUPAC Name5-bromo-2-fluorobenzonitrile
Molecular FormulaC7H3BrFN
InChI KeyGYCNHFWRPJXTSB-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1%
Assay (GC)≥97.5%
Melting Point (clear melt)77.0-83.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only