Mercury(II) iodide is is used for preparation of Nessler's reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mercury(II) iodide is is used for preparation of Nessler′s reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.
Solubility
Insoluble in water. Slightly soluble in alcohol, ether, acetone, chloroform, ethyl acetate, olive oil and castor oil.
Notes
Light sensitive. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Philip J.Potts; Peter C.Webb; Olwen Williams-Thorpe; Richard Kilworth. Analysis of silicate rocks using field-portable X-ray fluorescence instrumentation incorporating a mercury(II) iodide detector: a preliminary assessment of analytical performance. Analyst. 1995, 120, (5),1273-1278
- Ghodrat Mahmoudi; Ali Morsali. Mercury(II) iodide coordination polymers generated from polyimine ligands. Polyhedron. 2008, 27, (3),1070-1078