(+/-)-3-Chloro-1,2-propanediol, 98%
(+/-)-3-Chloro-1,2-propanediol, 98%
(+/-)-3-Chloro-1,2-propanediol, 98%
Thermo Scientific Chemicals

(+/-)-3-Chloro-1,2-propanediol, 98%

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2500 g
Catalog number A16143.36
also known as A16143-36
Price (EUR)
36,10
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
36,10
Each
Chemical Identifiers
CAS96-24-2
IUPAC Name3-chloropropane-1,2-diol
Molecular FormulaC3H7ClO2
InChI KeySSZWWUDQMAHNAQ-UHFFFAOYNA-N
SMILESOCC(O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4780-1.4820 @ 20*C
FormLiquid or viscous liquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only