Thermo Scientific Chemicals

L-(+)-Rhamnose monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16166.22
100 g, Each
Thermo Scientific Chemicals

L-(+)-Rhamnose monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16166.22
100 g, Each
Quantity
Catalog number: A16166.22
Price (USD)
Price: 316.00
Online price: 268.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10030-85-0
Water Content (Karl Fischer Titration)
5.2-14.13% (0.5-1.5 waters)
Optical Rotation
8.0 ± 1.0° (c=10 in water, 20hrs)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
88.0-97.0°C

Description

L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

Solubility
Soluble in water and methanol. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text