1,4-Diiodobutane, 99%, stab. with copper
1,4-Diiodobutane, 99%, stab. with copper
1,4-Diiodobutane, 99%, stab. with copper
Thermo Scientific Chemicals

1,4-Diiodobutane, 99%, stab. with copper

CAS: 628-21-7 | C4H8I2 | 309.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16216.22
also known as A16216-22
Price (EUR)
196,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
196,00
Each
Chemical Identifiers
CAS628-21-7
IUPAC Name1,4-diiodobutane
Molecular FormulaC4H8I2
InChI KeyROUYUBHVBIKMQO-UHFFFAOYSA-N
SMILESICCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow or pink
Refractive Index1.6195-1.6225 @ 20?C
Assay (GC)≥98.5%
StabilizerCopper powder
1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agent.
RUO – Research Use Only

General References:

  1. Lee, Y. H.; Park, J.; Lee, J.; Lee, S. U.; Lee, M. H. Iridium Cyclometalates with Tethered o-Carboranes: Impact of Restricted Rotation of o-Carborane on Phosphorescence Efficiency. J. Am. Chem. Soc. 2015, 137 (25), 8018-8021.
  2. Lyon, C. K.; Prasher, A.; Hanlon, A. M.; Tuten, B. T.; Tooley, C. A.; Frank, P. G.; Berda, E. B. A brief user's guide to single-chain nanoparticles. Polym. Chem. 2015, 6 (2), 181-197.