1,10-Phenanthroline monohydrochloride monohydrate, 99%
1,10-Phenanthroline monohydrochloride monohydrate, 99%
1,10-Phenanthroline monohydrochloride monohydrate, 99%
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrochloride monohydrate, 99%

CAS: 18851-33-7 | C12H11ClN2O | 234.683 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16254.22
also known as A16254-22
Price (USD)
500.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
500.00
Each
Chemical Identifiers
CAS18851-33-7
IUPAC Name1,10-phenanthroline hydrate hydrochloride
Molecular FormulaC12H11ClN2O
InChI KeyNDLHUHRGAIHALB-UHFFFAOYSA-N
SMILESO.Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay from Supplier's CofA≥98.5% (US-sourced material)
Identification (FTIR)Conforms (UK-sourced material)
CommentMaterial sourced in UK and US
Water Content (Karl Fischer Titration)7-10% (UK-sourced material)
View more
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
RUO – Research Use Only

General References:

  1. Liu, Y. L.; Xu, X. Y.; Sun, P. C.; Chen, T. H. N-doped porous carbon nanosheets with embedded iron carbide nanoparticles for oxygen reduction reaction in acidic media. Int. J. Hydrogen Energy 2015, 40 (13), 4531-4539.
  2. Vyprachticky, D.; Kankova, D.; Pokorna, V.; Kminek, I.; Dzhabarov, V.; Cimrova, V. Novel and Simple Synthesis of Brominated 1,10-Phenanthrolines. Aust. J. Chem. 2014, 67 (6), 915-921.