Di-n-butyl ether, 99%
Di-n-butyl ether, 99%
Di-n-butyl ether, 99%
Thermo Scientific Chemicals

Di-n-butyl ether, 99%

CAS: 142-96-1 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A16305.0F
also known as A16305-0F
Price (USD)
102.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
102.00
Each
Chemical Identifiers
CAS142-96-1
IUPAC Name1-butoxybutane
Molecular FormulaC8H18O
InChI KeyDURPTKYDGMDSBL-UHFFFAOYSA-N
SMILESCCCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.3970-1.4010 @ 20°C
Water Content≤0.05% (≤500ppm)
Di-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.

Solubility
Miscible with alcohol, ether and acetone. Immiscible with water.

Notes
Air and light sensitive. Incompatible with oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Pérez-Maciá, M. A.; Bringué, R.; Iborra, M.; Tejero, J.; Cunill, F. Thermodynamic equilibrium for the dehydration of 1-butanol to di-n-butyl ether. Chem. Eng. Res. Des. 2015, 102, 186-195.
  2. Cai, L.; Sudholt, A.; Lee, D. J.; Egolfopoulos, F. N.; Pitsch, H.; Westbrook, C. K.; Sarathy, S. M. Chemical kinetic study of a novel lignocellulosic biofuel: Di-n-butyl ether oxidation in a laminar flow reactor and flames. Combust. Flame 2014, 161 (3), 798-809.