Cobalt(II) chloride hexahydrate, 98%
Cobalt(II) chloride hexahydrate, 98%
Cobalt(II) chloride hexahydrate, 98%
Cobalt(II) chloride hexahydrate, 98%
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 98%

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16346.18
Price (USD)
32.80
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
32.80
Each
Add to cart
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-purple or purple
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (as hexahydrate)
Water Content (Karl Fischer Titration)42.2-48.4% (5.25-6.75 waters)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

Solubility
Soluble in water, ether, acetone and alcohol.

Notes
Incompatible with strong oxidizing agents and metals.
RUO – Research Use Only

General References:

  1. Kimer, J. T.; Elliott, C. M. Are High-Potential Cobalt Tris(bipyridyl) Complexes Sufficiently Stable to Be Efficient Mediators in Dye-Sensitized Solar Cells? Synthesis, Characterization, and Stability Tests. J. Phys. Chem. C 2015, 119 (31), 17502-17514.
  2. Parmentier, D.; Hoogerstraete, T. V.; Metz, S. J.; Binnemans, K.; Kroon, M. C. Selective Extraction of Metals from Chloride Solutions with the Tetraoctylphosphonium Oleate Ionic Liquid. Ind. Eng. Chem. Res. 2015, 54 (18), 5149-5158.