CD79a Antibody (24C2.5), FITC conjugate, Molecular Probes™

Catalog number: A16369

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Mouse
SubClass (Isotype): IgG1, kappa
Clonality: Monoclonal
Clone ID: 24C2.5
Antibody Fragment: Whole Antibody
Target Gene: CD79a
Gene ID: 12518
Gene Symbol: Cd79a
Gene Aliases: Ig-alpha, Iga, Igalpha, Ly-54, Ly54, mb-1
Label or Dye: FITC (Fluorescein)
Excitation⁄Emission (nm): 488⁄519
Excitation Class: Visible
Concentration: 0.5 mg⁄ml
Form: Liquid
Number of Reactions: 25 Tests
For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Size: 25 µg
Other Additives: Sodium Azide Preservative

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products