3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
el ácido 3,5-dinitrobenzoico se utiliza como reactivo en la derivación de resinas y la determinación de ampicilina.
Solubilidad
es soluble en etanol (50 mg/ml), metanol, alcohol (mucho) y ácido acético glacial (mucho). Insoluble en agua.
Notas
Almacenar en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Mantener alejado de agentes oxidantes fuertes, bases fuertes y agentes reductores fuertes.
RUO – Research Use Only
General References:
- VR Pedireddi.; S Varughese. Solvent-dependent coordination polymers: Cobalt complexes of 3, 5-dinitrobenzoic acid and3, 5-dinitro-4-methylbenzoic acid with 4, 4'-bipyrdine. Inorg. Chem. 2004, 43, (2), 450-457.
- JL Lopez.; FSG Einschlag.; MC González. Hydroxyl radical initiated photodegradation of 4-chloro-3, 5-dinitrobenzoic acid in aqueous solution. Journal of Photochemistry and Photobiology . 2000, 137, (2-3), 177-184.