What is the optimal passage number for reprogramming patient fibroblasts using the CytoTune-iPS 2.0 Sendai Reprogramming Kit?
We recommend reprogramming patient cells at the earliest passage possible. However, it is important to have the cells growing and healthy, which can take between 1-4 weeks. The cells are usually ready to reprogram once they have gone through a total of 3-4 passages.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How many cells do I need to start my reprogramming experiment with the CytoTune-iPS 2.0 Sendai Reprogramming Kit?
We recommend seeding approximately 50,000-100,000 fibroblast cells per well of a 6-well to yield 200,000-300,000 cells two days later. This equals a confluency of 50-80% on the day of transduction. Overly confluent cells will result in a decreased transduction efficiency. When cells are overly confluent (>80% confluent), we recommend re-plating the cells to achieve a range of 50-80% confluency on the day of transduction.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is the difference between the CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit and the CytoTune-iPS 2.0 Sendai Reprogramming Kit?
The CTS CytoTune-iPS 2.1 Sendai vector formulation does not contain the bovine serum albumin carrier protein. Additionally, the preparation contains no animal-derived components at the primary component level. Each virus (i.e., KOS, L-Myc, and Kfl4) comes in a volume of 200 µL instead of 100 µL. The titer of the CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit can range from 0.5 x 10E8 to 2.0 x 10E8 virus particles per mL whereas the titer of the CytoTune-iPS 2.0 Sendai Reprogramming Kit is typically around 1.0 x 10E8 virus particles per mL.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What do I do if my iPSCs still contain the Sendai virus after 10 passages?
The Invitrogen CytoTune-iPS Sendai 2.0 Reprogramming Kit contains a temperature sensitive mutant of c-Myc and KOS that facilitates the clearance of these vectors. To clear c-Myc and KOS, incubate the iPSCs at 38-39 degrees C for 5 days. One caveat is that given the sensitive nature of iPSCs, only perform this temperature shift if Sendai virus is in your iPSC lines after more than 10 passages, and you have performed RT-PCR to show that the Klf4 vector is absent from your cells (this vector does not have temperature-sensitive mutations). Then you can perform temperature shift to remove the c-Myc and KOS vectors.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
I am seeing cell death during reprogramming using the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit? Is this normal?
Depending on your cell type, you should expect to see some cytotoxicity 24-48 hours post-transduction, which can affect >50% of your cells. This is an indication of high uptake of the virus and is caused by the expression of exogenous genes. We recommend that you continue culturing your cells and proceed with the protocol.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is alkaline phosphatase (AP)?
AP is a phenotypic marker of pluripotent stem cells (PSCs), including undifferentiated embryonic stem cells (ESCs), induced pluripotent stem cells (iPSCs), embryonic germ cells (EGCs) and Embryonic Carcinoma Cells (ECCs). While AP is expressed in most cell types, its expression is highly elevated in PSCs. Therefore, AP staining has been used to differentially stain PSCs to easily distinguish them from mouse embryonic fibroblasts (MEFs) used as feeders and parental fibroblasts commonly used in reprogramming experiments.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Are the TaqMan Sendai Gene Expression assays compatible with all versions of CytoTune?
The TaqMan Sendai Gene Expression Assays are compatible with both CytoTune 2.0 and CytoTune 2.1, with one exception. The CytoTune 2.0 kit contains a vector which expresses c-Myc, while the CytoTune 2.1 kit contains a vector which expresses L-Myc. As such, the relevant assays should be used in conjunction with each kit (e.g. SEVCYMC for CytoTune 2.0, and SEVLMYC for CytoTune 2.1). All other assays are compatible with both kits.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Can I distinguish between the endogenous and exogenous expression of the genes (SOX2, Kfl4, cMyc, and Oct2)? Do you offer primers that are able to distinguish between endogenous and exogenous expression of these genes?
We do not have such primers for use in endpoint RT-PCR. However, we do offer validated TaqMan Gene Expression assays for each of the exogenous reprogramming genes (https://www.thermofisher.com/order/genome-database/browse/gene-expression/keyword/sendai+taqman). These assays are designed to specifically detect only expression of the exogenous genes found on the Sendai vectors, and will not cross-react with expression of the endogenous versions of these genes. Additionally, these assays can be used to detect the presence of each of the CytoTune Sendai vectors, for determination of clearance of the vectors from established iPSCs.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Which virus can be cleared faster by a temperature shift?
For the Invitrogen CytoTune 2.0 kit, which contains 3 vectors (KOS, cMyc, Klf4), both the cMyc and the KOS vector can be cleared by a temperature shift to 39 degrees C. The temperature shift will be most effective for the cMyc vector, but should still work for the KOS vector.
Note: Before performing a temperature shift, first verify that the individual Klf4 vector has cleared. The Klf4 vector will not be cleared by a temperature shift, and performing the shift while the Klf4 is still present could cause problems.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How do I confirm that my cells were transduced with the Sendai virus?
You can confirm the presence of the Sendai virus in your cells by several methods, including staining with anti-Sendai virus antibody, performing endpoint RT-PCR using the primer sets listed on page 50 in the CytoTune-iPS 2.0 Sendai Reprogramming Kit User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/cytotune_ips_2_0_sendai_reprog_kit_man.pdf), or real-time RT-PCR using the TaqMan Sendai Gene Expression assays, also listed on page 50 in the CytoTune-iPS 2.0 Sendai Reprogramming Kit User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/cytotune_ips_2_0_sendai_reprog_kit_man.pdf).
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How many passages does it take to clear the Invitrogen CytoTune 2.0 Sendai virus from the generated iPSCs?
It can take as few as five or as many as fifteen passages for the virus to clear from the cell. In rare cases, Sendai sequences can persist indefinitely. Clearance rate is clone-dependent and can be confirmed by PCR or by anti-Sendai antibody. For more information about generating vector-free iPSCs, please refer to the user manual (http://tools.thermofisher.com/content/sfs/manuals/cytotune_ips_2_0_sendai_reprog_kit_man.pdf).
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
The secondary antibody used in the live-cell staining protocol (both Invitrogen CytoTune and CytoTune 2.0 kits) is anti-IgG, whereas the primary antibody is IgM. Is there a typo, or can the anti-IgG secondary antibody recognize the IgM?
We have used this combination of primary and secondary with no problems. IgG secondary antibodies will cross-react with IgM primary antibodies, since IgM share the same kappa light chains as IgG.
The IgG secondary antibodies that are listed in manuals are available, and they will work against IgG primary antibodies, as well as IgM primary antibodies, and others. For that reason, we typically have those IgG secondary antibodies on hand, and use them in most applications. That is the main reason it was recommended.
Please note that the Invitrogen CytoTune-iPS Sendai Reprogramming Kit (Cat. Nos. A13780-01, A13780-02) has been discontinued.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How can I visualize the iPSCs?
The iPSC colonies can be easily visualized using alkaline phosphatase stain, such as the Alkaline Phosphatase Live Stain (Cat. No. A14353). In addition, reprogrammed colonies can be selected utilizing live staining with Tra1-60 or Tra1-81 antibodies that recognize undifferentiated iPSCs and enable the identification of reprogrammed cells from a variety of human cell types. Please refer to the user manual for the full protocol (http://tools.thermofisher.com/content/sfs/manuals/AP_Live_Stain_man.pdf).
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What protocol steps are needed if I am working with Invitrogen CytoTune 2.0 and want to reprogram PBMCs feeder-free and using Gibco Vitronectin?
If you want to reprogram PBMCs with Invitrogen CytoTune 2.0 under feeder-free conditions, you should follow the existing Invitrogen CytoTune 2.0 PBMC protocol, but plate onto Gibco Vitronectin or Geltrex matrix on day 3 instead of MEF, and then transition over to Gibco Essential 8 on Days 7-8 instead of Gibco KnockOut Serum Replacement-based PSC medium.
Efficiencies are typically lower than with feeder-dependent conditions, but you should still get some colonies.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
With the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit, how can I optimize my reprogramming efficiency?
We recommend first to try increasing the MOI of hKlf4 only. For example: go from 5:5:3 to 5:5:6. If optimization is still required, then increase the MOI of KOS and hc-Myc. The ratio of KOS and hc-Myc must be 1 to 1, and the MOI of hKlf4 can be varied independently. For example: go from 5:5:3 to 10:10:3 or 10:10:6.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
With the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit, how do I determine the volume of virus to add to my cells?
An MOI of 5:5:3 (KOS, hc-Myc, hKlf4) is recommended for most cell types. The virus titer varies from lot to lot; the required volume for each MOI is listed on the Certificate of Analysis (CoA) for each lot of product. You may also choose to optimize your MOI as this may vary depending on the cell type. The ratio of KOS and hc-Myc must be 1 to 1, and the MOI of hKlf4 can be varied independently. For example: if KOS is 4, then hc-Myc must also be 4.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
With the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit, what does "MOI of 5:5:3 (KOS, hc-Myc, hKlf4)" mean?
MOI (multiplicity of infection) describes the ratio of viral particles to cells. The three vectors in the Invitrogen CytoTune 2.0 Kit should each be added to the cells based on an MOI recommendation. We recommend the following MOIs as a starting point, and adjustments can be performed if reprogramming efficiency is not optimal:
- Invitrogen CytoTune 2.0 KOS: Starting MOI of 5
- Invitrogen CytoTune 2.0 hc-Myc: Starting MOI of 5
- Invitrogen CytoTune 2.0 hKlf4: Starting MOI of 3
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How many wells of a 6-well plate can I reprogram with one Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit?
One kit (one tube of each vector, i.e., 3 tubes) is sufficient for a minimum of 5 wells of a 6-well dish at MOI of 5:5:3 (KOS, hc-Myc, hKlf4) with a recommended plating density of 2 x 10E5 to 3 x 10E5 cells/well for human dermal fibroblasts. The virus can only be used once, as viral titers decrease significantly with each freeze-thaw cycle.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Are there safety concerns for Sendai (SeV) virus?
Although humans are not a natural host for SeV, and the virus is non-pathogenic to humans, appropriate care must be taken to prevent the potential mucosal exposure to the virus. The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit must be used under Biosafety Level 2 (BL-2) containment with biological safety cabinet and laminar flow hood, and with appropriate personal safety equipment to prevent mucosal exposure/splash.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What cell types have been successfully reprogrammed with Sendai virus?
Our reprogramming kits have been validated for a wide variety of cell types, including human fibroblasts, CD34+ cord blood cells, and peripheral blood mononuclear cells (PBMCs). For a current list of publications citing the cell types validated using this method, go here (http://www.thermofisher.com/us/en/home/life-science/stem-cell-research/induced-pluripotent-stem-cells/sendai-virus-reprogramming/cytotune-publications.html).
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What are the benefits of using an integration-free reprogramming method to generate iPSCs?
Integration-free reprogramming methods generate iPSCs that do not contain detectable vectors or transgenes. Traditional technologies used for reprogramming (e.g., lentivirus, retrovirus) integrate into the genome of the target cells. The resulting iPSCs and cells differentiated from those iPSCs will contain foreign DNA and could be unsafe and problematic for use in cell therapy and drug discovery applications. Furthermore, the integration could occur in a critical region of the genome, causing problems with unrelated developmental processes.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How does the Sendai virus (SeV) reprogram somatic cells?
The Sendai virus vectors in the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit are based on a modified, non-transmissible form of SeV, which has the fusion protein gene (F) deleted. The viral vectors maintain full infectivity to a wide range of cells; however they are no longer capable of producing infectious particles from infected cells because the viral genome lacks the F gene. The Sendai virus vectors contain transgenes that will express factors hOct3/4, hSox2, hKlf4, and hc-Myc. After transduction, the viral vectors will cause the cells to express these four genes, resulting in reprogramming
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is Sendai virus (SeV)?
Sendai virus, also known as Hemagglutinating Virus of Japan (HVJ), is a respiratory virus of mouse and rat first isolated in Sendai, Japan in the early 1950s. The virus is classified as mouse parainfluenza virus type I, belonging to the Paramyxoviridae family. SeV is an enveloped virus, 150-250 nm in diameter, whose genome is a single chain of (-) sense RNA (15,384 bases). The virus infects cells by attaching to the sialic acid receptor present on the surface of many different cells and is thus able to infect a wide range of cell types of various animal species.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How is the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit different from the original Invitrogen CytoTune-iPS Sendai Reprogramming Kit?
The Invitrogen CytoTune-iPS 2.0 Kit (Cat. Nos. A16517, A16518) offers higher reprogramming efficiency, faster clearance of the vectors, and lower cytotoxicity compared to the original Invitrogen CytoTune-iPS Sendai Reprogramming Kit (Cat. Nos. A13780-01, A13780-02) that has been discontinued.
The 2.0 kit contains three vectors, one of which is a polycistronic vector (Invitrogen CytoTune 2.0 KOS), designed to deliver increased reprogramming efficiency. This polycistronic vector has a different backbone containing temperature-sensitive mutations in the polymerase-related genes, and this helps to clear the virus faster after reprogramming and causes less cytotoxicity to the cells.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is the Invitrogen CytoTune-iPS Sendai Reprogramming Kit?
The Invitrogen CytoTune -iPS Sendai Reprogramming Kit (Cat. Nos. A13780-01, A13780-02) is a non-integrating system that reprograms somatic cells into induced pluripotent stem cells (iPSCs). This kit utilizes four Sendai virus-based vectors, each capable of expressing one of the four Yamanaka factors: Oct3/4, Sox2, Klf4, and c-Myc. The expression of these transcription factors in somatic cells has been shown to be a critical factor in the successful generation of iPSCs.
Please note that the Invitrogen CytoTune-iPS Sendai Reprogramming Kit (Cat. Nos. A13780-01, A13780-02) has been discontinued and replaced by the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit (Cat. Nos. A16517, A16518).
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Did you test the vector combinations that were chosen to make the Invitrogen CytoTune version 2 virus?
Yes we did, and found that only the combination of KOS (3 in 1), cMyc, and Klf4-vector yields highest reprogramming efficiency. For instance, KOS and cMyc alone are not sufficient for reprogramming. Addition of Oct4 or Sox2 results only in a very few reprogrammed colonies. This is mostly due to an imbalance in the stoichiometry of the reprogramming factors, which may impair the reprogramming efficiency significantly.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit?
The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit (Cat. Nos. A16517, A16518) is a non-integrating system that uses Sendai virus vectors to reprogram somatic cells into induced pluripotent stem cells (iPSCs). The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit contains three Invitrogen CytoTune 2.0 reprogramming vectors, including the four Yamanaka factors, Oct3/4, Sox2, Klf4, and c-Myc. The expression of these transcription factors in somatic cells has been shown to be a critical factor in the successful generation of iPSCs. Only one application of the vectors is required for successful reprogramming.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
When should I use the full length Human FGF-basic (FGF-2/bFGF) Recombinant Protein or the truncated variant, Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein?
The full length Human FGF-basic (FGF-2/bFGF) (aa 1-155) Recombinant Protein is recommended for stem cells whereas the truncated variant, Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein which is missing the first 9 amino acids, is recommended for use with neural and cardiac cells.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What are induced pluripotent stem cells?
Induced pluripotent stem cells (iPS or iPSCs) are pluripotent stem cells directly generated by introducing combination of genes coding for reprogramming factors into adult cells. These reprogramming factors include Oct4, Sox2, c-Myc, KLF4, NANOG, and LIN28. Yu, et al, generated iPS from a human mesenchymal cell line using lentiviral vectors carrying Oct4, Sox2, NANOG, and LIN28 genes (Science 318:1917 (2007)). Using a similar approach, Takahashi et al, generated iPS from human primary fibroblast cells by introducing genes coding for Oct3, Sox2, KLF4, and c-Myc into these cells (Cell 131:861 (2007)). iPS generated by reprogramming are similar to human ES cells in morphology, the capacity for unlimited proliferation, surface-antigen expression, gene expression, the ability to differentiate into cell types representing the three germ layers in vitro, and the ability to form teratomas after injection into SCID mice.