Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid
Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid
Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid
Thermo Scientific Chemicals

Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid

CAS: 3444-17-5 | C24H45CrO6 | 481.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A16560.30
also known as A16560-30
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS3444-17-5
IUPAC Namechromium(3+) tris(2-ethylhexanoate)
Molecular FormulaC24H45CrO6
InChI KeyWBKDDMYJLXVBNI-UHFFFAOYNA-K
SMILES[Cr+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Dark green viscous
Assay (unspecified)49 - 55% as salt by Cr Elemental Analysis
Elemental AnalysisCr 5.29 - 5.93%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only