Propargyl bromide, 80% in toluene, stab. with MgO
Propargyl bromide, 80% in toluene, stab. with MgO
Propargyl bromide, 80% in toluene, stab. with MgO
Thermo Scientific Chemicals

Propargyl bromide, 80% in toluene, stab. with MgO

CAS: 106-96-7 | C3H3Br | 118.961 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1000 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16580.AK
also known as A16580-AK
Price (EUR)
-
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS106-96-7
IUPAC Name3-bromoprop-1-yne
Molecular FormulaC3H3Br
InChI KeyYORCIIVHUBAYBQ-UHFFFAOYSA-N
SMILESBrCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay be hazy or contain sediment which is MgO stabilizer.
Refractive Index1.4910-1.4970 @ 20°C
Stabilizer0.3% MgO
Appearance (Color)Colorless to pale yellow to yellow-orange
FormLiquid
View more

Propargyl bromide, 80% in toluene is used to prepare betulonic acid-peptide conjugates with anti-inflammatory activity and propargylic amines employed in enyne metathesis. It finds application in the propargylation of spiro ketones, allylic alcohols and enone complexes. In Barbier-type reaction, it reacts with aldehydes to give alkyne alcohols. It is actively involved in the synthesis of polyethylene glycol (PEG) and peptide-grafted polyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl bromide, 80% in toluene is used to prepare betulonic acid-peptide conjugates with anti-inflammatory activity and propargylic amines employed in enyne metathesis. It finds application in the propargylation of spiro ketones, allylic alcohols and enone complexes. In Barbier-type reaction, it reacts with aldehydes to give alkyne alcohols. It is actively involved in the synthesis of polyethylene glycol (PEG) and peptide-grafted polyesters.

Solubility
Miscible with ethanol, ether, benzene, carbon tetrachloride and chloroform. Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible withbases, strong oxidizing agents, copper, peroxides, silver, silver oxides, mercury and mercury oxides.
RUO – Research Use Only

General References:

  1. Li, Y.; Cheng, B. One-pot synthesis of precise polyisoxazoles by click polymerization: Copper (I)-catalyzed 1, 3-dipolar cycloaddition of nitrile oxides with alkynes. J. Polym. Sci. A Polym. Chem. 2013, 51 (7), 1645-1650.
  2. Un, I.; Ibisoglu, H.; Un, S. S.; Cosut, B.; Kılıc, A. Syntheses, characterizations, thermal and photophysical properties of cyclophosphazenes containing adamantane units. Inorg. Chim. Acta 2013, 399, 219-226.