1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
el ácido 1,2,3,4-butanetracarboxílico se utiliza como agente de entrecruzamiento para preparar celulosa de algodón con el fin de mejorar las propiedades antiperla y pirorretardante. Se utiliza como espaciador en la unión entrecruzada de nanopartículas de titania con algodón. Además, se utiliza como agente de acabado de prensa duradero sin formaldehído.
Solubilidad
soluble en agua y etanol.
Notas
incompatible con agentes oxidantes fuertes.
RUO – Research Use Only
General References:
- Ji, B.; Tang, P.; Yan, K.; Sun, G. Catalytic actions of alkaline salts in reactions between 1,2,3,4-butanetetracarboxylic acid and cellulose: II. Esterification. Carbohydr. Polym. 2015, 132, 228-236.
- Kawase, T.; Nomura, Y.; Oida, T. Synthesis and Properties of 1,2,3,4-Butanetetracarboxylic Acid Type Gemini Surfactants with Semifluoroalkyl Chain as Hydrophobic Group. Tenside Surfact. Det. 2014, 52 (5), 386-395.