2-Chloro-4-methoxyphenol, 97%
2-Chloro-4-methoxyphenol, 97%
2-Chloro-4-methoxyphenol, 97%
2-Chloro-4-methoxyphenol, 97%
Thermo Scientific Chemicals

2-Chloro-4-methoxyphenol, 97%

CAS: 18113-03-6 | C7H7ClO2 | 158.581 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16652.14
also known as A16652-14
Price (EUR)
150,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
150,00
Each
Add to cart
Chemical Identifiers
CAS18113-03-6
IUPAC Name2-chloro-4-methoxyphenol
Molecular FormulaC7H7ClO2
InChI KeyGNVRRKLFFYSLGT-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
FormCrystalline powder or Fused solid
Assay (GC)≥96.0%
Melting Point (clear melt)36-44°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only