3-(Trifluoromethyl)benzyl alcohol, 97%
3-(Trifluoromethyl)benzyl alcohol, 97%
3-(Trifluoromethyl)benzyl alcohol, 97%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzyl alcohol, 97%

CAS: 349-75-7 | C8H7F3O | 176.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16739.14
also known as A16739-14
Price (EUR)
157,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
157,00
Each
Chemical Identifiers
CAS349-75-7
IUPAC Name[3-(trifluoromethyl)phenyl]methanol
Molecular FormulaC8H7F3O
InChI KeyBXEHKCUWIODEDE-UHFFFAOYSA-N
SMILESOCC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4575-1.4625 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only