2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
Thermo Scientific Chemicals

2,2':5',2″-Terthiophene, 99%

CAS: 1081-34-1 | C12H8S3 | 248.376 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number A16748.03
also known as A16748-03
Price (EUR)
231,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
231,00
Each
Chemical Identifiers
CAS1081-34-1
IUPAC Name5-(thiophen-2-yl)-2,2'-bithiophene
Molecular FormulaC12H8S3
InChI KeyKXSFECAJUBPPFE-UHFFFAOYSA-N
SMILESS1C=CC=C1C1=CC=C(S1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)90.0-96.0?C
2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Murat Ates; Nuri Eren. Electrochemical copolymerization of carbazole and 2,2':5'-2″ terthiophene: characterization and micro-capacitor application. Iranian Polymer Journal. 2014, 23 581-589.
  2. Mohammad Shamsuddin Ahmed; Haesang Jeong; Jung-Min You; Seungwon Jeon. Synthesis and characterization of an electrochromic copolymer based on 2,2':5',2″-terthiophene and 3,4-ethylenedioxythiophene. Applied Nanoscience. 2012, 2 133-141.