2-Mercaptopyridine N-oxide sodium salt, 40% aq. soln.
2-Mercaptopyridine N-oxide sodium salt, 40% aq. soln.
2-Mercaptopyridine N-oxide sodium salt, 40% aq. soln.
Thermo Scientific Chemicals

2-Mercaptopyridine N-oxide sodium salt, 40% aq. soln.

CAS: 3811-73-2 | C5H4NNaOS | 149.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A16922.AE
also known as A16922-AE
Price (EUR)
41,40
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
41,40
Each
Chemical Identifiers
CAS3811-73-2
IUPAC Namesodium 2-sulfanylidene-1,2-dihydropyridin-1-olate
Molecular FormulaC5H4NNaOS
InChI KeyXNRNJIIJLOFJEK-UHFFFAOYSA-N
SMILES[Na+].[O-]N1C=CC=CC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)39-43%
FormLiquid
Refractive Index1.4795- 1.4845 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptopyridine N-oxide sodium salt is one of the active components in paint, sealants, shampoo, adhesive and aerosol due to its anti-microbial activity. In biochemistry studies, it is utilized to transport zinc into cells. Further, it is used to form bidentate oxothiolane chelates with transition metals. It acts as a stabilizer and viscosity building provider in weak basic or neutral medium.

Solubility
Miscible with ethanol, propylene glycol, polyethylene glycol and dimethyl sulfoxide. Immiscible with liquid paraffin and olive oil.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Bretti, C.; Cigala, R. M.; Crea, F.; De Stefano, C.; Lando, G.; Sammartano, S. Thermodynamics of Zn2+ 2-mercaptopyridine-N-oxide and 2-hydroxypyridine-N-oxide interactions: Stability, solubility, activity coefficients and medium effects. J. Mol. Liq. 2015, 211, 876-884.
  2. Hassanpoor, S.; Khayatian, G. Combination of directly suspended droplet microextraction and flame atomic absorption spectrometry for determination of trace amounts of iron and copper. J. Braz. Chem. Soc. 2014, 25 (4), 734-742.
  3. Oonishi, Y. Development of Novel Cyclizations via Rhodacycle Intermediate and Its Application to Synthetic Organic Chemistry. Chem. Pharm. Bull. 2015, 63 (6), 397-407.