Thermo Scientific Chemicals

1-Bromo-2-iodobenzene, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A17020.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-2-iodobenzene, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A17020.06
5 g, Each
Quantity
Catalog number: A17020.06
also known as A17020-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
583-55-1
IUPAC Name
1-bromo-2-iodobenzene
Molecular Formula
C6H4BrI
InChI Key
OIRHKGBNGGSCGS-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=C1I
Appearance (Color)
Clear colorless to yellow to brown
Identification (FTIR)
Conforms
Refractive Index
1.6625-1.6675 @ 20?C
Assay (GC)
≥98.0%
Form
Liquid

Description

2-Bromoiodobenzene is used in the synthesis of diarylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoiodobenzene is used in the synthesis of diarylamines.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text