Tetraethyleneglycol monomethyl ether, 98%
Tetraethyleneglycol monomethyl ether, 98%
Tetraethyleneglycol monomethyl ether, 98%
Tetraethyleneglycol monomethyl ether, 98%
Thermo Scientific Chemicals

Tetraethyleneglycol monomethyl ether, 98%

CAS: 23783-42-8 | C9H20O5 | 208.254 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 A17021.06
亦稱為 A17021-06
價格 (TWD)
-
申請報價
Quantity:
5 g
Request bulk or custom format
化學識別
CAS23783-42-8
IUPAC Name2,5,8,11-tetraoxatridecan-13-ol
Molecular FormulaC9H20O5
InChI KeyZNYRFEPBTVGZDN-UHFFFAOYSA-N
SMILESCOCCOCCOCCOCCO
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4435-1.4475 @ 20?C
FormLiquid

A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

Solubility
Water solubility 999,999c.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Agustí Lledó and Julius Rebek Jr. Deep cavitand receptors with pH-independent water solubility.Chem. Commun., 2010,468630-8632.
  2. Bertrand Brizet.; Claire Bernhard.; Yulia Volkova.; Yoann Rousselin.; Pierre D. Harvey.; Christine Goze and Franck Denat. Boron functionalization of BODIPY by various alcohols and phenols.Org. Biomol. Chem.,2013,11,7729-7737.