DL-alpha-Tocopherol, 97+%
DL-alpha-Tocopherol, 97+%
DL-alpha-Tocopherol, 97+%
Thermo Scientific Chemicals

DL-alpha-Tocopherol, 97+%

CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
50 g
250 g
1000 g
產品號碼 A17039.18
亦稱為 A17039-18
價格 (TWD)
-
Quantity:
50 g
Request bulk or custom format
化學識別
CAS10191-41-0
IUPAC Name(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular FormulaC29H50O2
InChI KeyGVJHHUAWPYXKBD-IEOSBIPESA-N
SMILESCC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow or yellow to orange or yellow/brown
FormViscous liquid
Assay≥97.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Optical Rotation-0.02° to +0.02° (Neat, non-U.S. specification)
檢視更多
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

Solubility
Miscible with chloroform, vegetable oils,  ether, acetone and alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cimadevilla, H. M.; Hevia, D.; Miar, A.; Mayo, J. C.; Lombo, F.; Sainz, R. M. Development and validation of a single HPLC method for determination of alfa-tocopherol in cell culture and in human or mouse biological samples. Biomed. Chromatogr. 2015, 29 (6), 843-852.
  2. Powell, C. D.; Chowdhury, M. K.; Bureau, D. P. Assessing the bioavailability of l-lysine sulfate compared to l-lysine HCl in rainbow trout (Oncorhynchus mykiss). Aquacult. 2015, 448, (1), 327-333.