Di-n-butylgermanium dichloride, 97%
Di-n-butylgermanium dichloride, 97%
Di-n-butylgermanium dichloride, 97%
Thermo Scientific Chemicals

Di-n-butylgermanium dichloride, 97%

CAS: 4593-81-1 | C8H18Cl2Ge | 257.762 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17138.14
also known as A17138-14
Price (USD)
834.65
Special offer
Online exclusive
Ends: 30-Jun-2026
982.00
Save 147.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
834.65
Special offer
Online exclusive
Ends: 30-Jun-2026
982.00
Save 147.35 (15%)
Each
Chemical Identifiers
CAS4593-81-1
Molecular FormulaC8H22Cl2Ge
InChI KeyMRGRQNCCBMIDLM-UHFFFAOYSA-N
SMILESCl.Cl.CCCC[GeH2]CCCC
Molecular Weight (g/mol)261.79
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only