Thermo Scientific Chemicals

N-Carbamoyl-DL-aspartic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17166.18
50 g, Each
Thermo Scientific Chemicals

N-Carbamoyl-DL-aspartic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17166.18
50 g, Each
Quantity
Catalog number: A17166.18
also known as A17166-18
Price (USD)
Price: 1,508.00
Online price: 1,305.65
Your price:
Quantity
-

Chemical Identifiers

CAS
923-37-5
IUPAC Name
2-(carbamoylamino)butanedioic acid
Molecular Formula
C5H8N2O5
InChI Key
HLKXYZVTANABHZ-UHFFFAOYNA-N
SMILES
NC(=O)NC(CC(O)=O)C(O)=O
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

Solubility
Soluble in hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text