Nile Blue A
Nile Blue A
Nile Blue A
Thermo Scientific Chemicals

Nile Blue A

CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17174.14
also known as A17174-14
Price (EUR)
76,20
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
76,20
Each
Add to cart
Chemical Identifiers
CAS3625-57-8
IUPAC Name[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
Molecular FormulaC40H40N6O6S
InChI KeyQIRDPEPUXNCOLD-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark blue or purple or very dark brown
FormPowder
Loss on Drying≤8% (200?C)
lambda-max (UV-Vis)630-640nm (50% aq ethanol)
Specific Absorbance (E 1%/1cm)≥1500 at lambda max (50% aq ethanol)
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Jason D Slinker.; Natalie B Muren.; Sara E Renfrew.; Jacqueline K Barton. DNA charge transport over 34 nm.Nat Chem.2011,3(3), 228-233.
  2. Meikun Fan.; Penghui Wang.; Carlos Escobedo.; David Sinton.; Alexandre G Brolo. Surface-enhanced Raman scattering (SERS) optrodes for multiplexed on-chip sensing of nile blue A and oxazine 720.Lab Chip.2012,12,(8), 1554-1560 .