Thermo Scientific Chemicals

2-Bromopentane, tech. 90%, Thermo Scientific Chemicals

Catalog number: A17205.22
100 g, Each
Thermo Scientific Chemicals

2-Bromopentane, tech. 90%, Thermo Scientific Chemicals

Catalog number: A17205.22
100 g, Each
Quantity
Catalog number: A17205.22
also known as A17205-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
107-81-3
IUPAC Name
2-bromopentane
Molecular Formula
C5H11Br
InChI Key
LGAJYTCRJPCZRJ-UHFFFAOYNA-N
SMILES
CCCC(C)Br
Refractive Index
1.4365 - 1.4445
Form
liquid
Assay (GC)
> 88.0%
Appearance (Color)
Clear, colourless to pale yellow

Description

Used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chemical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text