3,3'-Thiodipropionic acid, 98%
3,3'-Thiodipropionic acid, 98%
3,3'-Thiodipropionic acid, 98%
Thermo Scientific Chemicals

3,3'-Thiodipropionic acid, 98%

CAS: 111-17-1 | C6H10O4S | 178.202 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17220.0B
also known as A17220-0B
Price (EUR)
53,60
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
53,60
Each
Chemical Identifiers
CAS111-17-1
IUPAC Name3-[(2-carboxyethyl)sulfanyl]propanoic acid
Molecular FormulaC6H10O4S
InChI KeyODJQKYXPKWQWNK-UHFFFAOYSA-N
SMILESOC(=O)CCSCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5% to ≤102.5% (Dry wt. basis)
3,3'-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

Solubility
Soluble in acetone, ethanol. Slightly soluble in water.

Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. S Bharathi.; V Yegnaraman.; GP Rao. Molecular recognition of alkali metal cations by a nickel hexacyanoferrate monolayer anchored to self-assembled monolayers of 3, 3'-thiodipropionic acid on gold. Langmuir.199511 (2), 666-668.
  2. N Bruland.; JH Wübbeler.; A Steinbüchel. 3-Mercaptopropionate Dioxygenase, a Cysteine Dioxygenase Homologue, Catalyzes the Initial Step of 3-Mercaptopropionate Catabolism in the 3,3-Thiodipropionic Acid-degrading BacteriumVariovorax paradoxus. The Journal of Biological Chemistry.2009284 (1), 660-672.