Thermo Scientific Chemicals

o-Xylylene dibromide, 97%, Thermo Scientific Chemicals

Catalog number: A17241.14
25 g, Each
Thermo Scientific Chemicals

o-Xylylene dibromide, 97%, Thermo Scientific Chemicals

Catalog number: A17241.14
25 g, Each
Quantity
Catalog number: A17241.14
Price (USD)
Price: 78.40
Online price: 66.65
Your price:
Quantity
-

Chemical Identifiers

CAS
91-13-4
IUPAC Name
1,2-bis(bromomethyl)benzene
Molecular Formula
C8H8Br2
InChI Key
KGKAYWMGPDWLQZ-UHFFFAOYSA-N
SMILES
BrCC1=CC=CC=C1CBr
Appearance (Color)
White to pale cream to pale orange to brown or pale pink
Assay (GC)
≥96.0% (non-U.S. specification)
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥96.0% (U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)

Description

1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

Solubility
Solubility in toluene is almost transparent and ethanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text