Bis(4-hydroxyphenyl) sulfone, 99%
Bis(4-hydroxyphenyl) sulfone, 99%
Bis(4-hydroxyphenyl) sulfone, 99%
Thermo Scientific Chemicals

Bis(4-hydroxyphenyl) sulfone, 99%

CAS: 80-09-1 | C12H10O4S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17342.30
also known as A17342-30
Price (EUR)
58,20
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
58,20
Each
Chemical Identifiers
CAS80-09-1
IUPAC Name4-(4-hydroxybenzenesulfonyl)phenol
Molecular FormulaC12H10O4S
InChI KeyVPWNQTHUCYMVMZ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)244.0-251.0?C
FormPowder
It is a reactant used in epoxy reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant used in epoxy reactions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Takashi Ikegami.; Takashi Mukawa.; Hiroyuki Nariai.; Toshifumi Takeuchia. Bisphenol A-recognition polymers prepared by covalent molecular imprinting. Anal. Chim. Acta. 2004, 504 (1),131-135.
  2. Hiroshi Ishibashi.; Naoko Watanabe.; Naomi Matsumura.; Masashi Hirano.; Yukiko Nagao.; Hideki Shiratsuchi.; Shinya Kohra.; Shin-ichi Yoshihara.; Koji Arizonoa. Toxicity to early life stages and an estrogenic effect of a bisphenol A metabolite, 4-methyl-2,4-bis(4-hydroxyphenyl)pent-1-ene on the medaka (Oryzias latipes). Life Sci.. 2005, 77 (21),2643-2655.