Amberlyst™ A-26(OH), ion exchange resin
Amberlyst™ A-26(OH), ion exchange resin
Amberlyst™ A-26(OH), ion exchange resin
Thermo Scientific Chemicals

Amberlyst™ A-26(OH), ion exchange resin

CAS: 39339-85-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17361.30
also known as A17361-30
Price (EUR)
76,65
Online Exclusive
85,40
Save 8,75 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
76,65
Online Exclusive
85,40
Save 8,75 (10%)
Each
Chemical Identifiers
CAS39339-85-0
MDL NumberMFCD00145579
SpecificationsSpecification SheetSpecification Sheet
FormBeads
CommentExchange capacity: ≥0.80 eq/L
Particle Size0.56-0.70mm
CommentMoisture holding capacity: 66-75% (OH form)
Appearance (Color)Moist pink or pale brown
Amberlyst™ A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

Solubility
Insoluble in water.

Notes
Exposure to air absorbs carbon dioxide. Incompatible with strong acids, bases and oxidants.
RUO – Research Use Only

General References:

  1. Macroreticular strongly basic anion excahnge resin, hydroxide form,suitable for aqueous and non-aqueous applications. Useful for removalof anionic metal complexes and mercaptans from non-polar solvents, andof acidic impurities from organic solutions.
  2. Haddad, R.; Thery, J.; Gauthier-Manuel, B.; Elouarzaki, K.; Holzinger, M.; Goff, A. L.; Gautier, G.; Mansouri, J. E.; Martinent, A.; Cosnier, S. High performance miniature glucose/O2 fuel cell based on porous silicon anion exchange membrane. Electrochem. Commun. 2015, 54, 10-13.
  3. Allen, C. E.; Curran, P. R.; Brearley, A. S.; Boissel, V.; Sviridenko, L.; Press, N. J.; Stonehouse, J. P.; Armstrong, A. Efficient and Facile Synthesis of Acrylamide Libraries for Protein-Guided Tethering. Org. Lett. 2015, 17 (3), 458-460.