Thermo Scientific Chemicals

p-Toluenesulfonyl isocyanate, 95%, Thermo Scientific Chemicals

Catalog number: A17492.36
500 g, Each
Thermo Scientific Chemicals

p-Toluenesulfonyl isocyanate, 95%, Thermo Scientific Chemicals

Catalog number: A17492.36
500 g, Each
Quantity
Catalog number: A17492.36
also known as A17492-36
Price (USD)
Price: 128.00
Online price: 108.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4083-64-1
Form
Liquid
Refractive Index
1.5320-1.5380 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms

Description

It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.

Solubility
Reacts with water.

Notes
Keep away from alcohols, strong bases, amines, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text