Diethyl phthalate, 99%
Diethyl phthalate, 99%
Diethyl phthalate, 99%
Diethyl phthalate, 99%
Thermo Scientific Chemicals

Diethyl phthalate, 99%

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
2500 g
Catalog number A17529.30
also known as A17529-30
Price (EUR)
24,60
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
24,60
Each
Add to cart
Chemical Identifiers
CAS84-66-2
IUPAC Name1,2-diethyl benzene-1,2-dicarboxylate
Molecular FormulaC12H14O4
InChI KeyFLKPEMZONWLCSK-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.5000-1.5035 @ 20?C
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
RUO – Research Use Only

General References:

  1. Wu, Y.; Si, Y.; Zhou, D.; Gao, J. Adsorption of diethyl phthalate ester to clay minerals. Chemosphere 2015, 119, 690-696.
  2. Fang, G.; Zhu, C.; Dionysiou, D. D.; Gao, J.; Zhou, D. Mechanism of hydroxyl radical generation from biochar suspensions: Implications to diethyl phthalate degradation. Bioresour. Technol. 2015, 176, 210-217.