1,9-Diphenyl-1,3,6,8-nonatetraen-5-one, 97+%, Thermo Scientific Chemicals
1,9-Diphenyl-1,3,6,8-nonatetraen-5-one, 97+%, Thermo Scientific Chemicals
1,9-Diphenyl-1,3,6,8-nonatetraen-5-one, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,9-Diphenyl-1,3,6,8-nonatetraen-5-one, 97+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A17568.06
also known as A17568-06
5 g
Catalog number A17568.06
also known as A17568-06
Price (EUR)
32,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
32,20
Each
Chemical Identifiers
CAS622-21-9
IUPAC Name(1E,3E,6E,8E)-1,9-diphenylnona-1,3,6,8-tetraen-5-one
Molecular FormulaC21H18O
InChI KeyRLJALOQFYHCJKG-FVRNMFRHSA-N
SMILESO=C(C=CC=CC1=CC=CC=C1)/C=C/C=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)137.0-146.0?C
Appearance (Color)Yellow
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only