Thermo Scientific Chemicals

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A17600.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A17600.14
25 g, Each
Quantity
Catalog number: A17600.14
also known as A17600-14
Price (USD)
Price: 132.00
Online price: 115.65
Your price:
Quantity
-

Chemical Identifiers

CAS
371-42-6
IUPAC Name
4-fluorobenzene-1-thiol
Molecular Formula
C6H5FS
InChI Key
OKIHXNKYYGUVTE-UHFFFAOYSA-N
SMILES
FC1=CC=C(S)C=C1
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5480-1.5520 @ 20?C

Description

It is used in the synthesis of Bicalutamide, an anti-cancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of Bicalutamide, an anti-cancer drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text