2,2'-Dibromobifenilo, 98 %, Thermo Scientific Chemicals
2,2'-Dibromobifenilo, 98 %, Thermo Scientific Chemicals
2,2'-Dibromobifenilo, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-Dibromobifenilo, 98 %, Thermo Scientific Chemicals

CAS: 13029-09-9 | C12H8Br2 | 312.004 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Número de catálogo A17632.14
también denominado A17632-14
Precio (EUR)
605,00
Each
Cantidad:
25 g
Pedido a granel o personalizado
Precio (EUR)
605,00
Each
Identificadores químicos
CAS13029-09-9
IUPAC Name2,2'-dibromo-1,1'-biphenyl
Molecular FormulaC12H8Br2
InChI KeyDRKHIWKXLZCAKP-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1=CC=CC=C1Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Melting Point (clear melt)76.0-83.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
2,2'-Dibromobiphenyl is used to produce 5,5-dimethyl-5H-dibenzosilole at the temperature of -78 - 20°C. It will need reagent n-BuLi and solvents diethyl ether, hexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
2,2′-dibromobifenilo se utiliza para producir 5,5-dimetil-5H-dibenzosilol a la temperatura de -78-20 °C. Necesitará reactivo n-BuLi y disolventes éter dietílico, hexano.

Solubilidad
Insoluble en agua.

Notas
Almacenar lejos de los agentes oxidantes fuertes. Mantener el recipiente herméticamente cerrado. Almacénese en un lugar fresco y seco en recipientes herméticos.
RUO – Research Use Only

General References:

  1. Herny Gilman, Bernard Gaj. Notes - Coupling Reactions with Some Organolithium Compounds in Tetrahydrofuran. J. Org. Chem. 1957, 22, (4), 447-449.
  2. M.D. Rausch.; G.A. Moser.; C.F. Meade. The isolation, characterization and synthetic utility of several solid organolithium compounds. Journal of Organometallic Chemistry. 1973, 51, 1-11.