2-Undecanone, 98%
2-Undecanone, 98%
2-Undecanone, 98%
Thermo Scientific Chemicals

2-Undecanone, 98%

CAS: 112-12-9 | C11H22O | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A17659.36
also known as A17659-36
Price (TWD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS112-12-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4275-1.4315 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only