Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A17719.36
500 g, Each
Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A17719.36
500 g, Each
Quantity
Catalog number: A17719.36
also known as A17719-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
19285-83-7
IUPAC Name
2-aminopentanedioic acid hydrate
Molecular Formula
C5H11NO5
InChI Key
OZDAOHVKBFBBMZ-UHFFFAOYNA-N
SMILES
O.NC(CCC(O)=O)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (dry wt. basis)
Optical Rotation
0° ±0.9° (c=1 in water)
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤15.0% (theory 10.91%)
Form
Crystalline powder or powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text