2-Nitrochalcone, 97%
2-Nitrochalcone, 97%
2-Nitrochalcone, 97%
Thermo Scientific Chemicals

2-Nitrochalcone, 97%

CAS: 7473-93-0 | C15H11NO3 | 253.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17763.14
also known as A17763-14
Price (EUR)
165,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS7473-93-0
IUPAC Name(2E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
Molecular FormulaC15H11NO3
InChI KeyKTXHLWZQKQDFRF-ZHACJKMWSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1C=CC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)121.0-127.0°C
Appearance (Color)Pale yellow to green
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only