4-Bromo-2,2-diphenylbutyronitrile, 95%
4-Bromo-2,2-diphenylbutyronitrile, 95%
4-Bromo-2,2-diphenylbutyronitrile, 95%
Thermo Scientific Chemicals

4-Bromo-2,2-diphenylbutyronitrile, 95%

CAS: 39186-58-8 | C16H14BrN | 300.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A17786.22
also known as A17786-22
Price (EUR)
108,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
108,00
Each
Chemical Identifiers
CAS39186-58-8
IUPAC Name4-bromo-2,2-diphenylbutanenitrile
Molecular FormulaC16H14BrN
InChI KeyIGYSFJHVFHNOEI-UHFFFAOYSA-N
SMILESBrCCC(C#N)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Melting Point (clear melt)62.0-75.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only